Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx3xb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130941.2 | NP_571016.2 | 8 - 9 | 1002 | 92 | |
XM_005168788.4 | XP_005168845.1 | 8 - 9 | 1103 | 92 | ||
XM_005168789.4 | XP_005168846.1 | 8 - 9 | 1103 | 92 | ||
XM_005168790.4 | XP_005168847.1 | 8 - 9 | 1055 | 92 | ||
XM_005168791.4 | XP_005168848.1 | 8 - 9 | 1103 | 92 | ||
XM_005168792.4 | XP_005168849.1 | 8 - 9 | 1103 | 92 | ||
GenBank mRNA | BC059794.1 | - | 8 - 9 | 1002 | 92 | |
Y12819.1 | - | 1007 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |