Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: six3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131363.1 | NP_571438.1 | 1 - 2 | 831 | 105 | |
GenBank mRNA | AB004881.1 | - | 1 - 2 | 897 | 105 | |
AF030281.1 | - | 1 - 2 | 831 | 105 | ||
BC059425.1 | - | 1 - 2 | 755 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |