Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp19a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131642.2 | NP_571717.2 | 6 - 7 | 837 | 102 | |
XM_021470523.1 | XP_021326198.1 | 6 - 7 | 1197 | 102 | ||
GenBank mRNA | AF183908.1 | - | 6 - 7 | 816 | 102 | |
AF226619.1 | - | 6 - 7 | 820 | 102 | ||
BC076104.1 | - | 7 - 8 | 741 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |