Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp1a2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131683.1 | NP_571758.1 | 20 - 21 | 2802 | 65 | |
GenBank mRNA | AF286373.1 | - | 20 - 21 | 2802 | 65 | |
AY028628.1 | - | 20 - 21 | 2736 | 65 | ||
BC163629.1 | - | 20 - 21 | 2724 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |