Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gad1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194419.1 | NP_919400.1 | 5 - 6 | 689 | 94 | |
XM_009302103.3 | XP_009300378.1 | 5 - 6 | 681 | 94 | ||
XM_009302104.3 | XP_009300379.1 | 4 - 5 | 866 | 94 | ||
GenBank mRNA | AB183390.1 | - | 5 - 6 | 873 | 94 | |
BC047851.1 | - | 5 - 6 | 689 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |