Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199713.2 | NP_956007.2 | 2 - 3 | 365 | 88 | |
GenBank mRNA | BC057469.1 | - | 2 - 3 | 303 | 88 | |
BC164724.1 | - | 2 - 3 | 230 | 88 | ||
EH505639.1 | - | 2 - 3 | 371 | 88 | ||
KM655668.1 | - | 2 - 3 | 196 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |