Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200394.1 | NP_956688.1 | 6 - 7 | 880 | 75 | |
GenBank mRNA | AY423041.1 | - | 6 - 7 | 807 | 75 | |
BC053411.1 | - | 6 - 7 | 880 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |