Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201034.2 | NP_957328.2 | 5 - 6 | 709 | 72 | |
GenBank mRNA | BC055159.1 | - | 5 - 6 | 654 | 72 | |
CT621066.2 | - | 5 - 6 | 568 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |