Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212565.1 | NP_997730.1 | 2 - 3 | 600 | 70 | |
GenBank mRNA | AY732218.2 | - | 2 - 3 | 425 | 70 | |
BC059592.1 | - | 2 - 3 | 600 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |