Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212685.1 | NP_997850.1 | 6 - 7 | 649 | 115 | |
GenBank mRNA | BC066624.1 | - | 6 - 7 | 649 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |