Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212892.1 | NP_998057.1 | 7 - 8 | 956 | 75 | |
GenBank mRNA | BC066616.1 | - | 7 - 8 | 956 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |