Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213426.1 | NP_998591.1 | 6 - 7 | 748 | 86 | |
GenBank mRNA | BC053413.1 | - | 6 - 7 | 748 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |