Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213494.2 | NP_998659.2 | 6 - 7 | 1246 | 68 | |
GenBank mRNA | BC055234.1 | - | 6 - 7 | 843 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |