Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli1rs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008780.1 | NP_001008780.1 | 4 - 5 | 703 | 108 | |
XM_005158200.3 | XP_005158257.1 | 4 - 5 | 779 | 108 | ||
XM_005158202.4 | XP_005158259.1 | 4 - 5 | 695 | 108 | ||
XM_005158203.4 | XP_005158260.1 | 4 - 5 | 695 | 108 | ||
XM_005158205.4 | XP_005158262.1 | 4 - 5 | 695 | 108 | ||
XM_009292528.3 | XP_009290803.1 | 4 - 5 | 811 | 108 | ||
XM_009292529.3 | XP_009290804.1 | 4 - 5 | 811 | 108 | ||
XM_009292530.3 | XP_009290805.1 | 4 - 5 | 810 | 108 | ||
XM_009292531.3 | XP_009290806.1 | 4 - 5 | 810 | 108 | ||
GenBank mRNA | AY839950.1 | - | 4 - 5 | 703 | 108 | |
BC055627.1 | - | 4 - 5 | 627 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |