Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020476.2 | NP_001018312.2 | 5 - 6 | 2233 | 93 | |
XM_005166992.4 | XP_005167049.1 | 5 - 6 | 2317 | 93 | ||
XM_005166993.4 | XP_005167050.1 | 5 - 6 | 2290 | 93 | ||
XM_005166994.4 | XP_005167051.1 | 5 - 6 | 2317 | 93 | ||
XM_005166996.4 | XP_005167053.1 | 5 - 6 | 2227 | 93 | ||
GenBank mRNA | AB196918.1 | - | 5 - 6 | 2233 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |