Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020476.2 | NP_001018312.2 | 10 - 11 | 2752 | 66 | |
XM_005166992.4 | XP_005167049.1 | 11 - 12 | 2968 | 66 | ||
XM_005166993.4 | XP_005167050.1 | 11 - 12 | 2941 | 66 | ||
XM_005166994.4 | XP_005167051.1 | 10 - 11 | 2899 | 66 | ||
XM_005166996.4 | XP_005167053.1 | 9 - 10 | 2677 | 66 | ||
GenBank mRNA | AB196918.1 | - | 9 - 10 | 2683 | 66 | |
BC163546.1 | - | 10 - 11 | 2744 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |