Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042688.2 | NP_001036153.2 | 2 - 3 | 1010 | 76 | |
GenBank mRNA | BC141795.1 | - | 2 - 3 | 1010 | 76 | |
DQ812118.1 | - | 2 - 3 | 681 | 76 | ||
KM655590.1 | - | 2 - 3 | 726 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |