Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045105.1 | NP_001038570.1 | 2 - 3 | 319 | 81 | |
XM_017357352.2 | XP_017212841.1 | 2 - 3 | 627 | 81 | ||
XM_021478287.1 | XP_021333962.1 | 2 - 3 | 368 | 81 | ||
GenBank mRNA | BC134845.1 | - | 2 - 3 | 230 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |