Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zglp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045449.1 | NP_001038914.1 | 3 - 4 | 1045 | 138 | |
XM_021476933.1 | XP_021332608.1 | 2 - 3 | 925 | 138 | ||
GenBank mRNA | BC122298.1 | - | 3 - 4 | 1045 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |