Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130936.1 | NP_571011.1 | 1 - 2 | 262 | 67 | |
GenBank mRNA | AF392996.1 | - | 1 - 2 | 262 | 67 | |
BC162384.1 | - | 1 - 2 | 92 | 67 | ||
BC162393.1 | - | 1 - 2 | 92 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |