Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jak2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131093.1 | NP_571168.1 | 3 - 4 | 346 | 74 | |
XM_009295057.3 | XP_009293332.1 | 3 - 4 | 524 | 74 | ||
GenBank mRNA | AJ005690.1 | - | 3 - 4 | 346 | 74 | |
BC162596.1 | - | 3 - 4 | 305 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |