Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131100.1 | NP_571175.1 | 1 - 2 | 153 | 54 | |
GenBank mRNA | AF071496.2 | - | 1 - 2 | 153 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |