Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131234.2 | NP_571309.2 | 4 - 5 | 938 | 63 | |
GenBank mRNA | BC124483.1 | - | 4 - 5 | 866 | 63 | |
BC164788.1 | - | 3 - 4 | 874 | 63 | ||
U18311.1 | - | 4 - 5 | 940 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |