Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131403.1 | NP_571478.1 | 2 - 3 | 1137 | 71 | |
XM_005157502.4 | XP_005157559.1 | 2 - 3 | 1446 | 71 | ||
GenBank mRNA | AF510736.1 | - | 2 - 3 | 1136 | 71 | |
BC056786.1 | - | 2 - 3 | 1137 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |