Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc43a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327986.1 | NP_001314915.1 | 4 - 5 | 693 | 136 | |
GenBank mRNA | BC044509.1 | - | 4 - 5 | 658 | 136 | |
BC164469.1 | - | 3 - 4 | 415 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |