Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200092.1 | NP_956386.1 | 7 - 8 | 813 | 74 | |
GenBank mRNA | BC049014.1 | - | 7 - 8 | 813 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |