Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapkapk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201303.1 | NP_958460.1 | 4 - 5 | 864 | 123 | |
GenBank mRNA | BC054572.1 | - | 4 - 5 | 864 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |