Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapkapk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201303.1 | NP_958460.1 | 6 - 7 | 1068 | 87 | |
GenBank mRNA | BC054572.1 | - | 6 - 7 | 1068 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |