Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp26b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212666.1 | NP_997831.1 | 2 - 3 | 633 | 83 | |
GenBank mRNA | AY321366.1 | - | 2 - 3 | 1181 | 83 | |
BC066759.1 | - | 2 - 3 | 633 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |