Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: th2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001829.1 | NP_001001829.1 | 1 - 2 | 303 | 66 | |
XM_005164734.4 | XP_005164791.1 | 1 - 2 | 631 | 66 | ||
GenBank mRNA | AY620823.1 | - | 1 - 2 | 303 | 66 | |
BC162637.1 | - | 1 - 2 | 298 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |