Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002579.1 | NP_001002579.1 | 2 - 3 | 298 | 61 | |
GenBank mRNA | AY847710.1 | - | 2 - 3 | 249 | 61 | |
BC076212.1 | - | 2 - 3 | 298 | 61 | ||
BC095310.1 | - | 2 - 3 | 251 | 61 | ||
BC165889.1 | - | 2 - 3 | 265 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |