Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003583.2 | NP_001003583.2 | 4 - 5 | 561 | 69 | |
XM_005172873.4 | XP_005172930.1 | 4 - 5 | 595 | 69 | ||
GenBank mRNA | BC078270.1 | - | 4 - 5 | 597 | 69 | |
CU468787.1 | - | 4 - 5 | 502 | 69 | ||
DQ224359.1 | - | 4 - 5 | 564 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |