Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nap1l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007453.1 | NP_001007454.1 | 6 - 7 | 555 | 122 | |
XM_009303210.3 | XP_009301485.1 | 6 - 7 | 632 | 122 | ||
XM_021477517.1 | XP_021333192.1 | 5 - 6 | 569 | 122 | ||
GenBank mRNA | BC085386.1 | - | 6 - 7 | 555 | 122 | |
BC164768.1 | - | 5 - 6 | 520 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |