Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif1al2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012371.2 | NP_001012371.2 | 7 - 8 | 974 | 74 | |
XM_005161350.4 | XP_005161407.1 | 8 - 9 | 1311 | 74 | ||
XM_021468817.1 | XP_021324492.1 | 9 - 10 | 1187 | 74 | ||
GenBank mRNA | AY835381.1 | - | 7 - 8 | 887 | 74 | |
BC093209.1 | - | 7 - 8 | 975 | 74 | ||
DT057411.1 | - | 2 - 3 | 261 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |