Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017816.1 | NP_001017816.1 | 2 - 3 | 502 | 126 | |
XM_009294455.3 | XP_009292730.1 | 2 - 3 | 544 | 126 | ||
GenBank mRNA | BC092802.1 | - | 2 - 3 | 502 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |