Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017885.1 | NP_001017885.1 | 7 - 8 | 997 | 120 | |
XM_021467652.1 | XP_021323327.1 | 6 - 7 | 1493 | 120 | ||
GenBank mRNA | BC092962.1 | - | 7 - 8 | 997 | 120 | |
BC164212.1 | - | 6 - 7 | 924 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |