Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044896.1 | NP_001038361.1 | 1 - 2 | 846 | 78 | |
XM_005168272.4 | XP_005168329.1 | 3 - 4 | 2551 | 78 | ||
XM_009295758.3 | XP_009294033.1 | 3 - 4 | 2223 | 78 | ||
GenBank mRNA | BC171546.1 | - | 1 - 2 | 884 | 78 | |
BC171548.1 | - | 1 - 2 | 884 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |