Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044896.1 | NP_001038361.1 | 2 - 3 | 1333 | 69 | |
XM_005168272.4 | XP_005168329.1 | 4 - 5 | 3038 | 69 | ||
XM_009295758.3 | XP_009294033.1 | 4 - 5 | 2710 | 69 | ||
GenBank mRNA | BC171546.1 | - | 2 - 3 | 1371 | 69 | |
BC171548.1 | - | 2 - 3 | 1371 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |