Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult2st3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078168.2 | NP_001071636.2 | 6 - 7 | 953 | 111 | |
XM_017351367.1 | XP_017206856.1 | 7 - 8 | 1024 | 111 | ||
GenBank mRNA | BC115350.1 | - | 6 - 7 | 953 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |