Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc17a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082835.1 | NP_001076304.1 | 2 - 3 | 357 | 101 | |
GenBank mRNA | EU873168.1 | - | 2 - 3 | 487 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |