Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082910.1 | NP_001076379.1 | 11 - 12 | 1165 | 144 | |
XM_009294060.3 | XP_009292335.1 | 11 - 12 | 1496 | 144 | ||
XM_017352282.2 | XP_017207771.1 | 10 - 11 | 1367 | 144 | ||
XM_017352285.2 | XP_017207774.1 | 3 - 4 | 272 | 144 | ||
XM_021468144.1 | XP_021323819.1 | 11 - 12 | 1268 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |