Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099447.1 | NP_001092917.1 | 2 - 3 | 435 | 94 | |
GenBank mRNA | BC171554.1 | - | 2 - 3 | 513 | 94 | |
BC171721.1 | - | 2 - 3 | 512 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |