Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131030.3 | NP_571105.3 | 6 - 7 | 758 | 83 | |
GenBank mRNA | BC049339.1 | - | 6 - 7 | 739 | 83 | |
U77627.1 | - | 6 - 7 | 724 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |