Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plrdgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131575.2 | NP_571650.2 | 17 - 18 | 2643 | 117 | |
GenBank mRNA | AF230807.1 | - | 17 - 18 | 2640 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |