Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153653.1 | NP_705939.1 | 1 - 2 | 160 | 93 | |
XM_021468749.1 | XP_021324424.1 | 1 - 2 | 299 | 93 | ||
GenBank mRNA | AF317643.1 | - | 1 - 2 | 160 | 93 | |
BC042317.1 | - | 1 - 2 | 293 | 93 | ||
BC066495.1 | - | 1 - 2 | 149 | 93 | ||
BC164717.1 | - | 1 - 2 | 176 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |