Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153653.1 | NP_705939.1 | 3 - 4 | 377 | 99 | |
XM_009295279.3 | XP_009293554.1 | 2 - 3 | 155 | 99 | ||
XM_021468749.1 | XP_021324424.1 | 3 - 4 | 516 | 99 | ||
GenBank mRNA | AF317643.1 | - | 3 - 4 | 377 | 99 | |
BC042317.1 | - | 3 - 4 | 537 | 99 | ||
BC066495.1 | - | 3 - 4 | 366 | 99 | ||
BC164717.1 | - | 3 - 4 | 420 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |