Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199732.2 | NP_956026.2 | 2 - 3 | 357 | 91 | |
GenBank mRNA | BC071509.1 | - | 2 - 3 | 357 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |