Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200649.1 | NP_956943.1 | 8 - 9 | 1319 | 97 | |
GenBank mRNA | BC057476.1 | - | 8 - 9 | 1319 | 97 | |
BC164569.1 | - | 7 - 8 | 1143 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |