Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpe65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200751.1 | NP_957045.1 | 4 - 5 | 389 | 68 | |
GenBank mRNA | AY646886.1 | - | 3 - 4 | 356 | 68 | |
BC059559.1 | - | 4 - 5 | 389 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |