Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nucb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201493.1 | NP_958901.1 | 4 - 5 | 490 | 92 | |
GenBank mRNA | BC046077.1 | - | 4 - 5 | 490 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |